5-Fluoro-2-methoxybenzyl bromide 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Bromomethyl)-4-fluoro-1-methoxybenzene

H25957-03 H25957-06
AAH25957-03EA 47.3 USD
AAH25957-03 AAH25957-06 AAH25957-14
5-Fluoro-2-methoxybenzyl bromide 98+%
5-Fluoro-2-methoxybenzyl bromide

Purity: 98+% CAS No.: 700381-18-6 Molecular Formula: C8H8BrFO Formula Weight: 219.05 Melting Point: 58-62° UN No.: 3261 MDL No.: MFCD00671769

Formula: C₈H₈BrFO
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00671769
CAS Number: 700381-18-6
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR