2-(4-Bromophenoxy)tetrahydropyran 98%
Supplier: Thermo Scientific Chemicals
H25914-18
H25914-09
AAH25914-18EA
156
USD
AAH25914-18
AAH25914-09
2-(4-Bromophenoxy)tetrahydropyran 98%
2-(4-Bromophenoxy)tetrahydropyran
Purity: 98%
CAS No.: 36603-49-3
Molecular Formula: C11H13BrO2
Formula Weight: 257.13
Melting Point: 55-58°
MDL No.: MFCD00091551
Formula:
C₁₁H₁₃BrO₂ Melting Pt: 55…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00091551 CAS Number: 36603-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...