3-Bromo-5-(trifluoromethyl)pyridine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H25823-06
H25823-03
AAH25823-06EA
361
USD
AAH25823-06
AAH25823-03
3-Bromo-5-(trifluoromethyl)pyridine 95%
3-Bromo-5-(trifluoromethyl)pyridine
Purity: 95%
CAS No.: 436799-33-6
Molecular Formula: C6H3BrF3N
Formula Weight: 226.00
Flash Point: 85°(185°F)
Melting Point: 44-46°
Boiling Point: 65-66°/19mm
MDL No.: MFCD04972700
Formula:
C₆H₃BrF₃N MW: 226.00 g/mol Boiling Pt: 65…66 °C/19 mm Melting Pt: 44…46 °C Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04972700 CAS Number: 436799-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...