3-Bromo-5-fluoropyridine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H25766-03
H25766-06
AAH25766-03EA
33.1
USD
AAH25766-03
AAH25766-06
3-Bromo-5-fluoropyridine 99%
3-Bromo-5-fluoropyridine
Purity: 99%
CAS No.: 407-20-5
Molecular Formula: C5H3BrFN
Formula Weight: 175.99
Flash Point: 64°(148°F)
Odor: Characteristic
UN No.: 2811
MDL No.: MFCD04112555
Formula:
C₅H₃BrFN MW: 175.99 g/mol Melting Pt: 24…28 °C Flash Pt: 64 °C(148 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04112555 CAS Number: 407-20-5 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...