2-Fluoro-5-(trifluoromethyl)phenyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Fluoro-5-(trifluoromethyl)phenylisocyanate
B25630-03
B25630-06
AAB25630-03EA
88.5
USD
AAB25630-03
AAB25630-06
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate 97%
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate
Purity: 97%
CAS No.: 69922-27-6
Molecular Formula: C8H3F4NO
Formula Weight: 205.11
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Refractive Index: 1.455
Boiling Point: 53°/2.5mm
UN No.: 2206
MDL No.: MFCD00673069
Formula:
C₈H₃F₄NO MW: 205.11 g/mol Boiling Pt: 53 °C (2.5 -rr) Density: 1.418 g/cm³ (20 °C) Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00673069 CAS Number: 69922-27-6 UN: 2206 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...