trans-Cinnamonitrile predominantly trans 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(E)-3-Phenylacrylonitrile, Cinnamonitrile
B25503-14
B25503-22
AAB25503-14EA
58.4
USD
AAB25503-14
AAB25503-22
AAB25503-36
trans-Cinnamonitrile predominantly trans 97%
trans-Cinnamonitrile
Purity: 97%
CAS No.: 1885-38-7
Molecular Formula: C9H7N
Formula Weight: 129.16
Flash Point: 126°(259°F)
Refractive Index: 1.6020
Melting Point: 18-20°
Boiling Point: 253-254°
MDL No.: MFCD00001930
Formula:
C₉H₇N MW: 129.16 g/mol Boiling Pt: 253…254 °C Melting Pt: 18…20 °C Density: 1.029 Flash Pt: 126 °C(259 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001930 CAS Number: 1885-38-7 EINECS: 217-552-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...