4-Fluoro-2-iodoaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25499-06
B25499-14
AAB25499-06EA
58
USD
AAB25499-06
AAB25499-14
4-Fluoro-2-iodoaniline 98%
4-Fluoro-2-iodoaniline
Purity: 98%
CAS No.: 61272-76-2
Molecular Formula: C6H5FIN
Formula Weight: 237.02
UN No.: 2810
MDL No.: MFCD00042584
Formula:
C₆H₅FIN MW: 237.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00042584 CAS Number: 61272-76-2 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...