Cyclopentanone oxime 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24961-14
B24961-22
AAAB24961-14EA
74.4
USD
AAAB24961-14
AAAB24961-22
Cyclopentanone oxime 97%
Cyclopentanone oxime
Purity: 97%
CAS No.: 1192-28-5
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 92°(198°F)
Melting Point: 54-58°
Boiling Point: 196°
MDL No.: MFCD00001420
Formula:
C₅H₉NO Boiling Pt: 196 °C Melting Pt: 54…58 °C Flash Pt: 92 °C (198 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001420 CAS Number: 1192-28-5 EINECS: 214-749-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...