Cobalt(II) thiocyanate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cobalt dirhodanide, Cobaltous rhodanide, Cobalt(II) thiocyanate, Cobalt dithiocyanate, Cobaltous thiocyanate

B23460-09 B23460-18 B23460-30
AAB23460-09EA 86.86 USD
AAB23460-09 AAB23460-18 AAB23460-30
Cobalt(II) thiocyanate 98+%
Cobalt (II) thiocyanate

Purity: 98+% CAS No.: 3017-60-5 Molecular Formula: C2CoN2S2 Formula Weight: 175.10 Odor: Odorless UN No.: 3077 MDL No.: MFCD00010937

Formula: C₂CoN₂S₂
Density: 1.955 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010937
CAS Number: 3017-60-5
EINECS: 221-156-8
UN: 3077
ADR: "9",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR