4-Formylmorpholine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Morpholine-4-carbaldehyde, N-Formylmorpholine
B23315-0E
B23315-22
B23315-36
AAB23315-0EEA
540
USD
AAB23315-0E
AAB23315-22
AAB23315-36
4-Formylmorpholine 99%
4-Formylmorpholine
Purity: 99%
CAS No.: 4394-85-8
Molecular Formula: C5H9NO2
Formula Weight: 115.13
Flash Point: 118°(244°F)
Refractive Index: 1.4860
Melting Point: 20-23°
Boiling Point: 239-241°
Beilstein Reference: 110293
MDL No.: MFCD00006170
Formula:
C₅H₉NO₂ MW: 115.13 g/mol Boiling Pt: 239…241 °C Melting Pt: 20…23 °C Density: 1.150 Flash Pt: 118 °C(244 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006170 CAS Number: 4394-85-8 EINECS: 224-518-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...