trans-2,5-Dimethylpiperazine 98%

Supplier: Thermo Scientific Chemicals
Danger

B23150-22 B23150-14 B23150-06
AAB23150-22EA 772 USD
AAB23150-22 AAB23150-14 AAB23150-06
trans-2,5-Dimethylpiperazine 98%
trans-2,5-Dimethylpiperazine

Purity: 98% CAS No.: 2815-34-1 Molecular Formula: C6H14N2 Formula Weight: 114.19 Flash Point: 58°(136°F) Melting Point: 115-119° Boiling Point: 162-165° Beilstein Reference: 79886 UN No.: 2925 MDL No.: MFCD00070516

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 162…165 °C
Melting Pt: 115…119 °C
Flash Pt: 58 °C(136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00070516
CAS Number: 2815-34-1
UN: 2925
ADR: "4.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR