2-Chloro-4-fluorophenylacetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(2-Chloro-4-fluorophenyl)acetonitrile

B23132-06 B23132-14
AAB23132-06EA 34 USD
AAB23132-06 AAB23132-14
2-Chloro-4-fluorophenylacetonitrile 97%
2-Chloro-4-fluorophenylacetonitrile

Purity: 97% CAS No.: 75279-56-0 Molecular Formula: C8H5ClFN Formula Weight: 169.59 Melting Point: 67-69° UN No.: 3439 MDL No.: MFCD00236029

Formula: C₈H₅ClFN
MW: 169.59 g/mol
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00236029
CAS Number: 75279-56-0
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR