Didodecyldimethylammonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning

B22839-04 B22839-09
AAB22839-04EA 47.6 USD
AAB22839-04 AAB22839-09
Didodecyldimethylammonium bromide 98+%
Didodecyldimethylammonium bromide

Purity: 98+% CAS No.: 3282-73-3 Molecular Formula: C26H56BrN Formula Weight: 462.65 Melting Point: 157-162° Beilstein Reference: 3576662

Formula: C₂₆H₅₆BrN
MW: 462.65 g/mol
Melting Pt: 157…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00041969
CAS Number: 3282-73-3
EINECS: 221-923-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR