Cobalt(II) chloride, anhydrous 97%

Supplier: Thermo Scientific Chemicals
Danger

B22031-14 B22031-22 B22031-36
AAB22031-14EA 71.34 USD
AAB22031-14 AAB22031-22 AAB22031-36
Cobalt(II) chloride, anhydrous 97%
Cobalt(II) chloride

Purity: 97% CAS No.: 7646-79-9 Molecular Formula: Cl2Co Formula Weight: 129.84 Odor: Odorless Melting Point: 735° Boiling Point: 1049° Merck Reference: 14,2437 UN No.: 3077 MDL No.: MFCD00010938

Formula: Cl₂Co
MW: 129.84 g/mol
Boiling Pt: 1049 °C (1013 hPa)
Melting Pt: 735 °C
Density: 1.6795 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010938
CAS Number: 7646-79-9
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 12,02498

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR