β-Chloro-4'-fluoropropiophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloro-1-(4-fluorophenyl)propan-1-one, 3-Chloro-4′-fluoropropiophenone, 3-Chloro-4'-fluoropropiophenone

B21891-22 B21891-14 B21891-06
AAAB21891-22EA 182 USD
AAAB21891-22 AAB21891-14 AAB21891-06
β-Chloro-4'-fluoropropiophenone 97%
β-Chloro-4'-fluoropropiophenone

Purity: 97% CAS No.: 347-93-3 Molecular Formula: C9H8ClFO Formula Weight: 186.61 Melting Point: 46-49° Boiling Point: 100-103°/3mm MDL No.: MFCD00000991

Formula: C₉H₈ClFO
MW: 186.61 g/mol
Boiling Pt: 100…103 °C/3 mm
Melting Pt: 46…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00000991
CAS Number: 347-93-3
EINECS: 206-475-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR