Diethyl tetradecylphosphonate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Diethyl 1-tetradecylphosphonate
B21426-06
B21426-14
AAB21426-06EA
80.1
USD
AAB21426-06
AAB21426-22
AAB21426-14
Diethyl tetradecylphosphonate 98%
Diethyl tetradecylphosphonate
Purity: 98%
CAS No.: 5191-09-3
Molecular Formula: C18H39O3P
Formula Weight: 334.47
Refractive Index: 1.4445
Melting Point: 8°
Boiling Point: 199-201°/3mm
MDL No.: MFCD00015597
Formula:
C₁₈H₃₉O₃P Boiling Pt: 199…201 °C/3 mm Melting Pt: 8 °C Density: 0.922 Storage Temperature: Ambient |
MDL Number:
MFCD00015597 CAS Number: 5191-09-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...