4-Aminomorpholine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Aminomorpholine
B21381-22
B21381-06
B21381-14
AAB21381-22EA
700
USD
AAB21381-22
AAB21381-06
AAB21381-14
4-Aminomorpholine 95%
4-Aminomorpholine
Purity: 98%
CAS No.: 4319-49-7
Molecular Formula: C4H10N2O
Formula Weight: 102.14
Storage Temperature: -30°C to -10°C
Flash Point: 58°(137°F)
Refractive Index: 1.4785
Boiling Point: 168°
UN No.: 2733
MDL No.: MFCD00006174
Formula:
C₄H₁₀N₂O Boiling Pt: 168 °C Density: 1.059 Flash Pt: 58 °C(137 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006174 CAS Number: 4319-49-7 EINECS: 224-347-4 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...