Triammonium hexafluoroaluminate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium hexafluoroaluminate

B20582-09 B20582-18
AAB20582-09EA 41.1 USD
AAB20582-09 AAB20582-18
Triammonium hexafluoroaluminate 98%
tri-Ammonium hexafluoroaluminate

Does not etch glass


Purity: 98% CAS No.: 7784-19-2 Molecular Formula: (NH4)3AlF6 Formula Weight: 195.09 Odor: Odorless Merck Reference: 14,524 MDL No.: MFCD00042144

Formula: H₁₂AlF₆N₃
MW: 195.09 g/mol
Density: 1.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042144
CAS Number: 7784-19-2
EINECS: 232-052-7
UN: 3288
ADR: 6.1,III
Merck Index: 12,00555

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR