Anthrone 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Anthracen-9(10H)-one

A19118-14 A19118-36 A19118-22
AAA19118-14EA 93 USD
AAA19118-14 AAAA19118-36 AAA19118-22
Anthrone 96%
Anthrone

Applications: In organic synthesis, in colorimetric determination of sugar in body fluids and of animal starch in liver tissue
Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water


Purity: 97% CAS No.: 90-44-8 Molecular Formula: C14H10O Formula Weight: 194.23 Melting Point: 153-160° Solubility: Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water Beilstein Reference: 1910173 Merck Reference: 14,691 MDL No.: MFCD00001187


In organic synthesis, in colorimetric determination of sugar in body fluids and of animal starch in liver tissue

Formula: C₁₄H₁₀O
MW: 194.23 g/mol
Melting Pt: 153…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00001187
CAS Number: 90-44-8
EINECS: 201-994-0
Merck Index: 14,691

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR