7-Amino-4-(trifluoromethyl)coumarin 99%

Supplier: Thermo Scientific Chemicals
Warning

A18400-01 A18400-03
AAA18400-01EA 41.4 USD
AAA18400-01 AAA18400-03
7-Amino-4-(trifluoromethyl)coumarin 99%
7-Amino-4-(trifluoromethyl)coumarin

Purity: 99% CAS No.: 53518-15-3 Molecular Formula: C10H6F3NO2 Formula Weight: 229.16 Melting Point: 222-223° Beilstein Reference: 4456797

Formula: C₁₀H₆F₃NO₂
MW: 229.16 g/mol
Melting Pt: 222…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00006858
CAS Number: 53518-15-3
EINECS: 258-599-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR