Cadmium(II) iodide 98%

Supplier: Thermo Scientific Chemicals
Danger

A18079-14 A18079-22
AAAA18079-14EA 48.7 USD
AAAA18079-14 AAAA18079-22
Cadmium(II) iodide 98%
Cadmium(II) iodide

Purity: 98% CAS No.: 7790-80-9 Molecular Formula: CdI2 Formula Weight: 366.21 Odor: Odorless Melting Point: 387-389° Boiling Point: 787° Merck Reference: 14,1621 UN No.: 2570 MDL No.: MFCD00010920

Formula: CdI₂
Boiling Pt: 787 °C
Melting Pt: 387…389 °C
Density: 5.67
Storage Temperature: Ambient
MDL Number: MFCD00010920
CAS Number: 7790-80-9
EINECS: 232-223-6
UN: 2570
ADR: 6.1,III
Merck Index: 14,1621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR