p-Tolyltrimethylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Trimethyl-p-tolylsilane, Trimethyl(p-tolyl)silane
A17884-06
A17884-14
AAA17884-06EA
96
USD
AAA17884-06
AAA17884-14
AAA17884-22
p-Tolyltrimethylsilane 97%
p-Tolyltrimethylsilane
Purity: 97%
CAS No.: 3728-43-6
Molecular Formula: C10H16Si
Formula Weight: 164.33
Flash Point: 46°(115°F)
Refractive Index: 1.4910
Boiling Point: 191-192°
UN No.: 1993
MDL No.: MFCD00014850
Formula:
C₁₀H₁₆Si MW: 164.32 g/mol Boiling Pt: 191…192 °C Density: 0.865 Flash Pt: 46 °C(115 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014850 CAS Number: 3728-43-6 UN: 1993 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...