Ethylphosphonic dichloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A17869-06 A17869-14
AAAA17869-06EA 88.8 USD
AAAA17869-06 AAAA17869-14
Ethylphosphonic dichloride 98%
Ethylphosphonic dichloride

Purity: 98% CAS No.: 1066-50-8 Molecular Formula: C2H5Cl2OP Formula Weight: 146.94 Flash Point: 110°(230°F) Refractive Index: 1.4665 Boiling Point: 71-72°/12mm UN No.: 2922 MDL No.: MFCD00002073

Formula: C₂H₅Cl₂OP
Boiling Pt: 71…72 °C/12 mm
Density: 1.376
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002073
CAS Number: 1066-50-8
EINECS: 213-918-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR