Cyclopentanecarbonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

A17710-03 A17710-06 A17710-14
AAA17710-03EA 62.1 USD
AAA17710-03 AAA17710-06 AAA17710-14
Cyclopentanecarbonitrile 98%
Cyclopentanecarbonitrile

Purity: 98% CAS No.: 4254-02-8 Molecular Formula: C6H9N Formula Weight: 95.14 Flash Point: 56°(133°F) Refractive Index: 1.4410 Melting Point: -76° Boiling Point: 67-68°/10mm Beilstein Reference: 1098717 UN No.: 1992 MDL No.: MFCD00013741

Formula: C₆H₉N
MW: 95.14 g/mol
Boiling Pt: 67…68 °C/10 mm
Melting Pt: –76 °C
Density: 0.912
Flash Pt: 56 °C (133 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013741
CAS Number: 4254-02-8
EINECS: 224-229-2
UN: 1992
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR