Tetradecylphosphonic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Tetradecylphosphonic acid, n-Tetradecylphosphonic acid, TDPA, N-tetradecylphosphonicacid
A17511-03
A17511-14
A17511-06
AAAA17511-03EA
107
USD
AAAA17511-03
AAAA17511-14
AAAA17511-06
Tetradecylphosphonic acid 98%
Tetradecylphosphonic acid
Purity: 98%
CAS No.: 4671-75-4
Molecular Formula: CH3(CH2)13P(O)(OH)2
Formula Weight: 278.37
MDL No.: MFCD00015834
Formula:
C₁₄H₃₁O₃P MW: 278.37 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00015834 CAS Number: 4671-75-4 EINECS: 225-118-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...