Bismuth antimonide 99%

Supplier: Thermo Scientific Chemicals
Warning

A17014-09 A17014-18
AAAA17014-09EA 59.9 USD
AAAA17014-09 AAAA17014-18
Bismuth antimonide 99%
Bismuth antimonide

Purity: 99.99% CAS No.: 12323-19-2 Molecular Formula: BiSb Formula Weight: 330.73 UN No.: 1549 MDL No.: MFCD00014196

Formula: BiSb
Storage Temperature: Ambient
MDL Number: MFCD00014196
CAS Number: 12323-19-2
UN: 1549
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR