Triethanolamine hydrochloride 99+%
Supplier: Thermo Scientific Chemicals
A15678-0B
A15678-0I
A15678-30
AAA15678-0BEA
161
USD
AAA15678-0B
AAA15678-0I
AAA15678-30
Triethanolamine hydrochloride 99+%
Triethanolamine hydrochloride
Protein purification reagent
Purity: 99+%
CAS No.: 637-39-8
Molecular Formula: C6H15NO3.HCl
Formula Weight: 185.65
Melting Point: 177-179°
Beilstein Reference: 3909940
Merck Reference: 14,9665
MDL No.: MFCD00012596
Formula:
C₆H₁₆ClNO₃ MW: 185.65 g/mol Melting Pt: 177…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012596 CAS Number: 637-39-8 EINECS: 211-284-2 Merck Index: 14,9665 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...