Tetraheptylammonium bromide 99%

Supplier: Thermo Scientific Chemicals
Warning

A15218-22 A15218-06 A15218-14
AAA15218-22EA 413 USD
AAA15218-22 AAAA15218-06 AAAA15218-14
Tetraheptylammonium bromide 99%
Tetraheptylammonium bromide

Purity: 99% CAS No.: 4368-51-8 Molecular Formula: [CH3(CH2)6]4NBr Formula Weight: 490.70 Melting Point: 87-89° MDL No.: MFCD00011861

Formula: C₂₈H₆₀BrN
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00011861
CAS Number: 4368-51-8
EINECS: 224-459-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR