(Iodomethyl)trimethylsilane 99%

Supplier: Thermo Scientific Chemicals
Warning

A15120-09 A15120-18
AAA15120-09EA 103 USD
AAA15120-09 AAA15120-18
(Iodomethyl)trimethylsilane 99%
(Iodomethyl)trimethylsilane

Purity: 99% CAS No.: 4206-67-1 Molecular Formula: C4H11ISi Formula Weight: 214.12 Flash Point: 31°(88°F) Refractive Index: 1.4915 Odor: Recognizable Boiling Point: 140-142° Beilstein Reference: 1731485 UN No.: 1993 MDL No.: MFCD00001077

Formula: C₄H₁₁ISi
MW: 214.12 g/mol
Boiling Pt: 140…142 °C
Density: 1.4301 g/cm³ (20 °C)
Flash Pt: 31 °C (88 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001077
CAS Number: 4206-67-1
EINECS: 224-123-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR