Methyl phenyl sulfoxide 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
(Methylsulphinyl)benzene
A15009-06
A15009-14
A15009-22
AAA15009-06EA
46.9
USD
AAA15009-06
AAA15009-14
AAA15009-22
Methyl phenyl sulfoxide 98+%
Methyl phenyl sulphoxide
Purity: 98+%
CAS No.: 1193-82-4
Molecular Formula: C7H8OS
Formula Weight: 140.20
Storage Temperature: 2°C to 8°C
Flash Point: 86°(186°F)
Refractive Index: 1.5775
Melting Point: 26-32°
Boiling Point: 93-94°/0.3mm
Beilstein Reference: 1904976
MDL No.: MFCD00002088
Formula:
C₇H₈OS MW: 140.21 g/mol Boiling Pt: 93…94 °C/0.3 mm Melting Pt: 26…32 °C Flash Pt: 86 °C(186 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002088 CAS Number: 1193-82-4 EINECS: 214-781-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...