Phenylarsine oxide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
PAO, Oxophenylarsine, Arzene
A14674-03
A14674-06
A14674-14
AAAA14674-03EA
45.24
USD
AAAA14674-03
AAAA14674-06
AAAA14674-14
Phenylarsine oxide 97%
Phenylarsine oxide
Purity: 97%
CAS No.: 637-03-6
Molecular Formula: C6H5AsO
Formula Weight: 168.03
Melting Point: 140-160°
Beilstein Reference: 2935227
Merck Reference: 14,6949
UN No.: 3465
MDL No.: MFCD00001990
Formula:
C₆H₅AsO Melting Pt: 140…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001990 CAS Number: 637-03-6 EINECS: 211-275-3 UN: 3465 ADR: "6.1",III Merck Index: 14,6949 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...