Homophthalic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Carboxymethyl)benzoic acid

A14297-18 A14297-30
AAA14297-18EA 69.66 USD
AAA14297-18 AAA14297-30
Homophthalic acid 99%
Homophthalic acid

Purity: 99% CAS No.: 89-51-0 Molecular Formula: C9H8O4 Formula Weight: 180.16 Melting Point: ca 180° dec. Beilstein Reference: 1872069

Formula: C₉H₈O₄
Melting Pt: ca 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004326
CAS Number: 89-51-0
EINECS: 201-913-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR