Dimethyl phenylphosphonite 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Phenyldimethoxyphosphine

A14006-06 A14006-03 A14006-14
AAA14006-06EA 85.1 USD
AAA14006-06 AAAA14006-03 AAA14006-14
Dimethyl phenylphosphonite 98%
Dimethyl phenylphosphonite

Purity: 98% CAS No.: 2946-61-4 Molecular Formula: C6H5P(OCH3)2 Formula Weight: 170.15 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5290 Beilstein Reference: 1073018 MDL No.: MFCD00008352

Formula: C₈H₁₁O₂P
MW: 170.15 g/mol
Density: 1.072
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008352
CAS Number: 2946-61-4
EINECS: 220-960-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR