4-Chlorobenzylamine 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(4-Chlorophenyl)methanamine
A13984-06
A13984-14
A13984-22
AAA13984-06EA
31.5
USD
AAA13984-06
AAA13984-14
AAA13984-22
4-Chlorobenzylamine 97+%
4-Chlorobenzylamine
Purity: 97+%
CAS No.: 104-86-9
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Flash Point: 90°(194°F)
Refractive Index: 1.5590
Odor: Amine-like
Boiling Point: 229°
Beilstein Reference: 507949
UN No.: 2735
MDL No.: MFCD00008121
Formula:
C₇H₈ClN MW: 141.6 g/mol Boiling Pt: 229 °C Density: 1.160 Flash Pt: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008121 CAS Number: 104-86-9 EINECS: 203-245-3 UN: 2922 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...