Ammonium carbonate ≥30% (NH₃ basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Salt of hartshorn

A13955-0B A13955-0I
AAAA13955-0BEA 71.6 USD
AAAA13955-0B AAAA13955-0I
Ammonium carbonate ≥30% (NH₃ basis)
Ammonium carbonate

CAS No.: 506-87-6 Molecular Formula: CH8N2O3 Formula Weight: 96.09 Odor: Ammonia-like Melting Point: 60° subl. Beilstein Reference: 3627235 Merck Reference: 14,508 MDL No.: MFCD00010890

Formula: (NH₄)₂CO₃
MW: 96.09 g/mol
Melting Pt: 58 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR