Oil red O
Supplier: Thermo Scientific Chemicals
Synonyms:
CI 26125, Sudan Red 5B, Solvent Red 27
A12989-14
A12989-22
AAA12989-14EA
45.76
USD
AAA12989-14
AAA12989-22
Oil red O
Oil Red O
Suitable as a lipid and lipoprotein stain on cellulose acetate
CAS No.: 1320-06-5
Molecular Formula: C26H24N4O
Formula Weight: 408.51
Melting Point: ca 166° dec.
Beilstein Reference: 3491382
MDL No.: MFCD00003898
Formula:
C₂₆H₂₄N₄O MW: 408.51 g/mol Melting Pt: ca 166 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003898 CAS Number: 1320-06-5 EINECS: 215-295-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...