Diisopropyl methylphosphonate 95%

Supplier: Thermo Scientific Chemicals
Warning

A12904-22 A12904-06 A12904-14
AAAA12904-22EA 694 USD
AAAA12904-22 AAAA12904-06 AAAA12904-14
Diisopropyl methylphosphonate 95%
Diisopropyl methylphosphonate

Purity: 95% CAS No.: 1445-75-6 Molecular Formula: C7H17O3P Formula Weight: 180.18 Refractive Index: 1.4100 Boiling Point: 50-51°/1mm MDL No.: MFCD00015022

Formula: C₇H₁₇O₃P
MW: 180.18 g/mol
Boiling Pt: 50…51 °C/1 mm
Density: 0.976
Storage Temperature: Ambient
MDL Number: MFCD00015022
CAS Number: 1445-75-6
EINECS: 215-896-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR