4-Ethylpyrocatechol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Ethylbenzene-1,2-diol, 4-Ethylcatechol

A12048-MD A12048-03 A12048-06
AAAA12048-MDEA 42 USD
AAAA12048-MD AAA12048-03 AAA12048-06
4-Ethylpyrocatechol 98%
4-Ethylpyrocatechol

Purity: 98% CAS No.: 1124-39-6 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 37-40° Boiling Point: 105-107°/0.5mm MDL No.: MFCD00015847

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 105…107 °C (0.5 mmHg)
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00015847
CAS Number: 1124-39-6
EINECS: 214-397-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR