Deoxybenzoin 97%

Supplier: Thermo Scientific Chemicals

A10580-14 A10580-22
AAA10580-14EA 42.1 USD
AAA10580-14 AAA10580-22
Deoxybenzoin 97%
Deoxybenzoin

Purity: 97% CAS No.: 451-40-1 Molecular Formula: C14H12O Formula Weight: 196.25 Flash Point: 110°(230°F) Melting Point: 53-57° Boiling Point: 319-321° Beilstein Reference: 1072876 MDL No.: MFCD00003081

Formula: C₁₄H₁₂O
Boiling Pt: 319…321 °C
Melting Pt: 53…57 °C
Density: 1.2
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003081
CAS Number: 451-40-1
EINECS: 207-193-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR