Phenyl dichlorophosphate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phenyl phosphorodichloridate
A10479-14
A10479-36
A10479-22
AAA10479-14EA
31.1
USD
AAA10479-14
AAA10479-36
AAA10479-22
Phenyl dichlorophosphate 97%
Phenyl dichlorophosphate
Reagent for preparation of phosphate diesters
Purity: 97%
CAS No.: 770-12-7
Molecular Formula: C6H5Cl2O2P
Formula Weight: 210.98
Flash Point: 112°(233°F)
Refractive Index: 1.5230
Melting Point: -1°
Boiling Point: 241-243°
Beilstein Reference: 511863
UN No.: 3265
MDL No.: MFCD00002067
Formula:
C₆H₅Cl₂O₂P MW: 210.98 g/mol Boiling Pt: 241…243 °C Melting Pt: –1 °C Density: 1.415 Flash Pt: 112 °C (233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002067 CAS Number: 770-12-7 EINECS: 212-220-6 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...