Cyclobutanecarbonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

A10309-14 A10309-06
AAA10309-22EA 2325.63 USD
AAA10309-22 AAA10309-14 AAA10309-06
Cyclobutanecarbonitrile 97%
Cyclobutanecarbonitrile

Purity: 97% CAS No.: 4426-11-3 Molecular Formula: C5H7N Formula Weight: 81.12 Flash Point: 42°(107°F) Refractive Index: 1.4325 Odor: Recognizable Boiling Point: 144-146° Beilstein Reference: 2037573 UN No.: 1992 MDL No.: MFCD00013735

Formula: C₅H₇N
MW: 81.12 g/mol
Boiling Pt: 144…146 °C
Density: 0.868
Flash Pt: 42 °C(107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013735
CAS Number: 4426-11-3
EINECS: 224-606-1
UN: 1992
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR