Ytterbium ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
045920-04
045920-09
045920-18
AA45920-04EA
144
USD
AA45920-04
AA45920-09
AA45920-18
Ytterbium ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh
Ytterbium
Purity: 99.9%
CAS No.: 7440-64-4
Physical Form: Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Silver
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00011286
Formula:
Yb Storage Temperature: Ambient |
MDL Number:
MFCD00011286 CAS Number: 7440-64-4 EINECS: 231-173-2 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...