Silver ≥99.9% (metals basis), wire 0.404 mm (0.0159 in) Ø, annealed, Diameter 0.404 mm (0.0159 in)

Supplier: Thermo Scientific Chemicals

045852-G9 045852-H4
AA45852-G9EA 365 USD
AA45852-G9 AA45852-H4
Silver ≥99.9% (metals basis), wire 0.404 mm (0.0159 in) Ø, annealed, Diameter 0.404 mm (0.0159 in)
Silver

Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈1.35g/m Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR