Dicopper selenide ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Copper(I) selenide
045660-09
045660-18
AA45660-09EA
92.03
USD
AA45660-09
AA45660-18
Dicopper selenide ≥99.5% (metals basis)
Dicopper selenide
Purity: 99.5%
CAS No.: 20405-64-5
Molecular Formula: Cu2Se
Formula Weight: 206.05
Physical Form: 2mm & down pieces
Density: 6.84
Appearance: Dark blue to black
Melting Point: 1113°
Merck Reference: 14,2666
UN No.: 3077
MDL No.: MFCD00049908
Formula:
Cu₂Se MW: 206.05 g/mol Melting Pt: 1113 °C Density: 6.84 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049908 CAS Number: 20405-64-5 EINECS: 243-796-7 UN: 3077 ADR: 9,III Merck Index: 12,02736 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...