Hafnium(IV) chloride ≥99.9% (metals basis) (max. 0.5% Zr)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hafnium tetrachloride

045612-14 045612-22 045612-36
AA45612-14EA 121.28 USD
AA45612-14 AA45612-22 AA45612-36
Hafnium(IV) chloride ≥99.9% (metals basis) (max. 0.5% Zr)
Hafnium(IV) chloride

Purity: 99.9% CAS No.: 13499-05-3 Molecular Formula: Cl4Hf Formula Weight: 320.30 Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: White Odor: Irritating Melting Point: 320° subl. Merck Reference: 14,4588 UN No.: 3260 MDL No.: MFCD00011034

Formula: Cl₄Hf
MW: 320.3 g/mol
Melting Pt: 431.85 (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00011034
CAS Number: 13499-05-3
EINECS: 236-826-5
UN: 3260
ADR: 8 II
Merck Index: 14,4588

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR