Digallium triselenide ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gallium (III) selenide

045572-03 045572-06
AA45572-03EA 48.3 USD
AA45572-03 AA45572-06
Digallium triselenide ≥99.99% (metals basis)
Digallium triselenide

Purity: 99.99% CAS No.: 12024-24-7 Molecular Formula: Ga2Se3 Formula Weight: 376.33 Physical Form: 3-12mm Sintered Lump UN No.: 3077 MDL No.: MFCD00016103

Formula: Ga₂Se₃
MW: 376.33 g/mol
Melting Pt: 1010…1030 °C
Density: 4.92 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016103
CAS Number: 12024-24-7
EINECS: 234-693-8
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR