Dirhenium heptaoxide ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Rhenium (VII) oxide
045538-04
045538-09
AA45538-04EA
189
USD
AA45538-04
AA45538-09
Dirhenium heptaoxide ≥99.995% (metals basis)
Dirhenium heptaoxide
Purity: 99.995%
CAS No.: 1314-68-7
Molecular Formula: O7Re2
Formula Weight: 484.40
Physical Form: Crystalline, Ampouled under argon
Density: 6.103
Appearance: Yellow to pale green
Odor: Odorless
Melting Point: 297°
Boiling Point: 360°
Merck Reference: 14,8178
UN No.: 3085
MDL No.: MFCD00011199
Formula:
O₇Re₂ MW: 484.41 g/mol Boiling Pt: 360 °C Melting Pt: 297 °C Density: 6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011199 CAS Number: 1314-68-7 EINECS: 215-241-9 UN: 3085 ADR: 5.1,III Merck Index: 12,08344 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...