Arsenic (III) sulfide ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Arsenic trisulphide, Disrsenic trisulphide
045522-06
045522-14
AA45522-06EA
141
USD
AA45522-06
AA45522-14
Arsenic (III) sulfide ≥99.999% (metals basis)
Arsenic(III) sulphide
Purity: 99.999%
CAS No.: 1303-33-9
Molecular Formula: As2S3
Formula Weight: 246.03
Physical Form: -100 Mesh Powder
Density: 3.43
Appearance: Yellow
Melting Point: 300°
Boiling Point: 707°
Merck Reference: 14,806
UN No.: 1557
MDL No.: MFCD00003435
Formula:
As₂S₃ MW: 246.04 g/mol Boiling Pt: 707 °C Melting Pt: 300 °C Density: 4.25 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003435 CAS Number: 1303-33-9 EINECS: 215-117-4 UN: 1557 ADR: 6.1,III Merck Index: 12,00846 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...