Decafluoro-4,6-bis(trifluoromethyl)cyclohexane tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Perfluoro-1,3-dimethylcyclohexane, Perfluoro(1,3-dimethylcyclohexane)
045477-18
045477-30
AA45477-18EA
70.5
USD
AA45477-18
AA45477-30
Decafluoro-4,6-bis(trifluoromethyl)cyclohexane tech. 90%, Technical Grade
Decafluoro-4,6-bis(trifluoromethyl)cyclohexane
Purity: 90%
CAS No.: 335-27-3
Molecular Formula: C8F16
Formula Weight: 400.06
Physical Form: Liquid
Density: 1.828
Melting Point: -55°
Boiling Point: 101-102°
Beilstein Reference: 2064147
MDL No.: MFCD00001469
Formula:
C₈F₁₆ MW: 400.06 g/mol Boiling Pt: 101…102 °C Melting Pt: –55 °C Density: 1.828 Storage Temperature: Ambient |
MDL Number:
MFCD00001469 CAS Number: 335-27-3 EINECS: 206-386-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...