Antimony (III) sulfide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Needle antimony, Antimony(III) sulphide, Amtimony sulphide, Diantimony trisulphide, Stibnite, Antimony glance, Antimony trisulphide, Antimonous sulphide, Antimony sulphide black

045464-09 045464-18
AA45464-09EA 115 USD
AA45464-09 AA45464-18
Antimony (III) sulfide ≥99.999% (metals basis)
Antimony (III) sulphide

Purity: 99.999% CAS No.: 1345-04-6 Molecular Formula: Sb2S3 Formula Weight: 339.68 Physical Form: 6mm & down pieces Appearance: Black Melting Point: 550° Merck Reference: 14,713 UN No.: 1549 MDL No.: MFCD00011217

Formula: S₃Sb₂
MW: 339.72 g/mol
Boiling Pt: 1150 °C (1013 hPa)
Melting Pt: 550 °C
Density: 4.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011217
CAS Number: 1345-04-6
EINECS: 215-713-4
UN: 1549
ADR: 6.1,III
Merck Index: 12,00754

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR